Anti-NOX1 Rabbit Polyclonal Antibody
Catalog # 77737-481
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y5S8
- Antigen Synonyms:GP91 2|Mitogenic oxidase (pyridine nucleotide dependent superoxide generating)|Mitogenic oxidase 1|MOX-1|MOX1|NADH/NADPH mitogenic oxidase subunit P65 MOX|NADH/NADPH mitogenic oxidase subunit P65-MOX|NADPH oxidase 1|NADPH oxidase 1 variant NOH 1L|NADPH oxidase homolog 1|NOH 1|NOH-1|NOH1|NOX-1|NOX1_HUMAN|RP1 146H21.1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:GP91 2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA:1:5000
Type:
Antigen: NOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse