Anti-HAVCR2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: HAVCR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-444
- 77856-741
- Antigen Symbol:
- HAVCR2
- HAVCR2
- Antigen Synonyms:
- CD366|FLJ14428|HAVcr-2|HAVR2_HUMAN|Hepatitis A virus cellular receptor 2|Kidney injury molecule 3|KIM 3|KIM3|T cell immunoglobulin and mucin domain containing 3|T cell immunoglobulin mucin 3|T-cell immunoglobulin and mucin domain-containing protein 3|T-cell immunoglobulin mucin family member 3|T-cell immunoglobulin mucin receptor 3|T-cell membrane protein 3|Tim 3|TIM-3|TIM3|TIMD-3|TIMD3
- CD366|FLJ14428|HAVcr-2|HAVR2_HUMAN|Hepatitis A virus cellular receptor 2|Kidney injury molecule 3|KIM 3|KIM3|T cell immunoglobulin and mucin domain containing 3|T cell immunoglobulin mucin 3|T-cell immunoglobulin and mucin domain-containing protein 3|T-cell immunoglobulin mucin family member 3|T-cell immunoglobulin mucin receptor 3|T-cell membrane protein 3|Tim 3|TIM-3|TIM3|TIMD-3|TIMD3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDQ0
- Q8TDQ0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Hepatitis A virus cellular receptor 2 protein (22-202AA)
- Recombinant human Hepatitis A virus cellular receptor 2 protein (22-202AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together