Anti-MAT2A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAT2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-513
- 77742-816
- Antigen Symbol:
- MAT2A
- MAT2A
- Antigen Synonyms:
- AdoMet synthase 2|AdoMet synthetase 2|AdoMet synthetase|AMS 2|AMS2|MAT 2|MAT 2A|MAT II|MAT-II|MATA 2|MATA2|MATII|Methionine adenosyltransferase 2|Methionine adenosyltransferase|Methionine adenosyltransferase II alpha|Methionine adenosyltransferase II|METK2_HUMAN|S adenosylmethionine synthetase gamma form|S adenosylmethionine synthetase isoform type 2|S-adenosylmethionine synthase isoform type-2|SAMS 2|SAMS2
- AdoMet synthase 2|AdoMet synthetase 2|AdoMet synthetase|AMS 2|AMS2|MAT 2|MAT 2A|MAT II|MAT-II|MATA 2|MATA2|MATII|Methionine adenosyltransferase 2|Methionine adenosyltransferase|Methionine adenosyltransferase II alpha|Methionine adenosyltransferase II|METK2_HUMAN|S adenosylmethionine synthetase gamma form|S adenosylmethionine synthetase isoform type 2|S-adenosylmethionine synthase isoform type-2|SAMS 2|SAMS2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31153
- P31153
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human S-adenosylmethionine synthase isoform type-2 protein (1-395AA)
- Recombinant Human S-adenosylmethionine synthase isoform type-2 protein (1-395AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together