Anti-KDR/FLT4 Rabbit Polyclonal Antibody
77738-651
New Product
- Antigen Symbol:KDR/FLT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P35968/P35916
- Antigen Synonyms:VEGFR2|VEGFR3|FLT4|CD antigen CD309|FLK1|Fetal liver kinase 1|Vascular endothelial growth|Protein-tyrosine kinase receptor flk-1|VEGFR-2|FLK-1|KDR|Vascular endothelial growth factor receptor 2|Kinase insert domain receptor
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:KDR
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:40000
Type:
Antigen: KDR/FLT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat