Anti-GPD2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-306
- 77738-742
- Antigen Symbol:
- GPD2
- GPD2
- Antigen Synonyms:
- GDH2|Glycerol 3 phosphate dehydrogenase 2 (mitochondrial)|Glycerol 3 phosphate dehydrogenase 2|Glycerol 3 phosphate dehydrogenase mitochondrial|Glycerol-3-phosphate dehydrogenase|GPD 2|GPD M|GPD-M|GPDH M|GPDH-M|GPDM|GPDM_HUMAN|mGPDH|mitochondrial|Mitochondrial glycerophosphate dehydrogenase|mtGPD|OTTHUMP00000162788|OTTHUMP00000204452|OTTHUMP00000204453|OTTHUMP00000204458
- GDH2|Glycerol 3 phosphate dehydrogenase 2 (mitochondrial)|Glycerol 3 phosphate dehydrogenase 2|Glycerol 3 phosphate dehydrogenase mitochondrial|Glycerol-3-phosphate dehydrogenase|GPD 2|GPD M|GPD-M|GPDH M|GPDH-M|GPDM|GPDM_HUMAN|mGPDH|mitochondrial|Mitochondrial glycerophosphate dehydrogenase|mtGPD|OTTHUMP00000162788|OTTHUMP00000204452|OTTHUMP00000204453|OTTHUMP00000204458
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43304
- P43304
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Glycerol-3-phosphate dehydrogenase, mitochondrial protein (43-727AA)
- Recombinant human Glycerol-3-phosphate dehydrogenase, mitochondrial protein (43-727AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together