Anti-RHOBTB3 Rabbit Polyclonal Antibody
77736-698
New Product
- Antigen Symbol:RHOBTB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O94955
- Antigen Synonyms:RHOBTB3|KIAA0878|Rho-related BTB domain-containing protein 3|EC 3.6.1.-
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:EC 3.6.1.-
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, IHC:1:100 to 1:300, IHC:1:200 to 1:1000, ELISA:1:40000
Type:
Antigen: RHOBTB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse