Anti-MRE11 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MRE11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-372
- 77859-935
- Antigen Symbol:
- MRE11
- MRE11
- Antigen Synonyms:
- meiotic recombination (S. cerevisiae) 11 homolog A|MRE11 homolog A|MRE11_HUMAN|MRE11b|OTTHUMP00000236833|OTTHUMP00000236831|OTTHUMP00000236832|OTTHUMP00000236830|AT like disease|MRE11 homolog 1|Mre 11|MRE11 meiotic recombination 11 homolog A (S. cerevisiae)|Double-strand break repair protein MRE11A|HNGS1|Meiotic recombination 11 homolog 1|DNA recombination and repair protein|MRE 11a|MRE11A|Ataxia telangiectasia disorder like|MRE 11b|MmMRE11A|endo/exonuclease MRE11 homolog double strand break repair nuclease|MRE11 meiotic recombination 11 homolog A|meiotic recombination 11 homolog A (S. cerevisiae)|Meiotic recombination 11 homolog A|ATLD|Double strand break repair protein MRE11A
- AT like disease|Ataxia telangiectasia disorder like|ATLD|DNA recombination and repair protein|Double strand break repair protein MRE11A|Double-strand break repair protein MRE11A|endo/exonuclease HNGS1|meiotic recombination (S. cerevisiae) 11 homolog A|Meiotic recombination 11 homolog 1|meiotic recombination 11 homolog A (S. cerevisiae)|Meiotic recombination 11 homolog A|MmMRE11A|Mre 11|MRE 11a|MRE 11b|MRE11 homolog 1|MRE11 homolog A|MRE11 homolog double strand break repair nuclease|MRE11 meiotic recombination 11 homolog A (S. cerevisiae)|MRE11 meiotic recombination 11 homolog A|MRE11_HUMAN|MRE11A|MRE11b|OTTHUMP00000236830|OTTHUMP00000236831|OTTHUMP00000236832|OTTHUMP00000236833
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49959
- P49959
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Double-strand break repair protein MRE11A protein (1-205AA)
- Recombinant Human Double-strand break repair protein MRE11A protein (1-205AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together