Anti-ITK Rabbit Polyclonal Antibody
77740-845
New Product
- Antigen Symbol:ITK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q08881
- Antigen Synonyms:EMT|Homolog of mouse T cell itk/tsk|IL 2 inducible T cell kinase|IL2 inducible T cell kinase|Interleukin 2 inducible T cell kinase|Itk|ITK_HUMAN|Kinase EMT|LPFS1|LYK|MGC126257|MGC126258|PSCTK 2|PSCTK2|T cell specific kinase|T-cell-specific kinase|TSK|Tyrosine protein kinase|Tyrosine protein kinase ITK/TSK|Tyrosine protein kinase Lyk|Tyrosine-protein kinase ITK/TSK|Tyrosine-protein kinase Lyk
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:EMT
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: ITK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse