Anti-GPR37 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: GPR37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-360
- 77738-867
- Antigen Symbol:
- GPR37
- GPR37
- Antigen Synonyms:
- EDNRBL| Endothelin B receptor like protein 1| Endothelin B receptor-like protein 1| Endothelin receptor type B like| ETBR LP 1| ETBR-LP-1| ETBRLP1| G protein coupled receptor 37 | G protein coupled receptor 37 endothelin receptor type B like| Gpr37| GPR37_HUMAN| hET(B)R LP| hET(B)RLP| PAELR| Parkin associated endothelin receptor like receptor | Parkin-associated endothelin receptor-like receptor| Probable G protein coupled receptor 37 | Probable G-protein coupled receptor 37
- EDNRBL|Endothelin B receptor like protein 1|Endothelin B receptor-like protein 1|Endothelin receptor type B like|ETBR LP 1|ETBR-LP-1|ETBRLP1|G protein coupled receptor 37 |G protein coupled receptor 37 endothelin receptor type B like|Gpr37|GPR37_HUMAN|hET(B)R LP|hET(B)RLP|PAELR|Parkin associated endothelin receptor like receptor |Parkin-associated endothelin receptor-like receptor|Probable G protein coupled receptor 37 |Probable G-protein coupled receptor 37
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15354
- O15354
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Prosaposin receptor GPR37 protein (114-133AA)
- Peptide sequence from Human Prosaposin receptor GPR37 protein (114-133AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together