Anti-ATP6V1F Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: ATP6V1F
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-363
- 77744-364
- Antigen Symbol:
- ATP6V1F
- ATP6V1F
- Antigen Synonyms:
- Adenosinetriphosphatase 14k chain| ATP6S14| ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F| ATPase, vacuolar, 14 kD| H(+)-transporting two-sector ATPase, 14kD subunit| MGC117321| MGC126037| MGC126038| V-ATPase 14 kDa subunit| V-ATPase subunit F| V-type proton ATPase subunit F| Vacuolar ATP synthase subunit F| Vacuolar proton pump subunit F| VATF| VATF_HUMAN| Vma7
- Adenosinetriphosphatase 14k chain| ATP6S14| ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F| ATPase, vacuolar, 14 kD| H(+)-transporting two-sector ATPase, 14kD subunit| MGC117321| MGC126037| MGC126038| V-ATPase 14 kDa subunit| V-ATPase subunit F| V-type proton ATPase subunit F| Vacuolar ATP synthase subunit F| Vacuolar proton pump subunit F| VATF| VATF_HUMAN| Vma7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16864
- Q16864
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- V-type proton ATPase subunit F protein (1-119AA)
- V-type proton ATPase subunit F protein (1-119AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together