Anti-GHRH Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GHRH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Specifications
- Catalog No:
- 77738-238
- 77855-981
- Antigen Symbol:
- GHRH
- GHRH
- Antigen Synonyms:
- Somatoliberin|Growth hormone-releasing factor|GRF|Growth hormone-releasing hormone
- Somatoliberin|Growth hormone-releasing factor|GRF|Growth hormone-releasing hormone
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Sheep
- Sheep
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07217
- P07217
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Sheep Somatoliberin protein (1-44AA)
- Recombinant Sheep somatoliberin protein (1-44AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together