Anti-NUF2 Rabbit Polyclonal Antibody
Catalog # 77745-181
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NUF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9BZD4
- Antigen Synonyms:Cancer/testis antigen 106|CDCA 1|CDCA1|Cell division cycle associated 1|Cell division cycle associated protein 1|Cell division cycle-associated protein 1|CT106|hNuf 2|hNuf2|hNuf2R|hsNuf2|Kinetochore protein Nuf2|Nuf 2|NUF2|NUF2 NDC80 kinetochore complex component|NUF2 NDC80 kinetochore complex component homolog|NUF2, S. cerevisiae, homolog of|NUF2_HUMAN|NUF2R
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:NUF2R
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NUF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat