Anti-FGFR2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FGFR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-162
- 77855-389
- Antigen Symbol:
- FGFR2
- FGFR2
- Antigen Synonyms:
- bacteria-expressed kinase|BBDS|BEK|BEK fibroblast growth factor receptor|BFR1|CD332|CD332 antigen|CEK3|CFD1|Craniofacial dysostosis 1|ECT1|FGF receptor|FGFR 2|FGFR-2|Fgfr2|FGFR2_HUMAN|Fibroblast growth factor receptor 2|Hydroxyaryl protein kinase|Jackson Weiss syndrome|JWS|K SAM|K-sam|Keratinocyte growth factor receptor 2|Keratinocyte growth factor receptor|KGFR|KSAM|protein tyrosine kinase|receptor like 14|soluble FGFR4 variant 4|TK14|TK25
- bacteria-expressed kinase|BBDS|BEK|BEK fibroblast growth factor receptor|BFR1|CD332|CD332 antigen|CEK3|CFD1|Craniofacial dysostosis 1|ECT1|FGF receptor|FGFR 2|FGFR-2|Fgfr2|FGFR2_HUMAN|Fibroblast growth factor receptor 2|Hydroxyaryl protein kinase|Jackson Weiss syndrome|JWS|K SAM|K-sam|Keratinocyte growth factor receptor 2|Keratinocyte growth factor receptor|KGFR|KSAM|protein tyrosine kinase|receptor like 14|soluble FGFR4 variant 4|TK14|TK25
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21802
- P21802
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fibroblast growth factor receptor 2 protein (577-821AA)
- Recombinant Human Fibroblast growth factor receptor 2 protein (577-821AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together