Anti-ESRP2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ESRP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-197
- 77854-939
- Antigen Symbol:
- ESRP2
- ESRP2
- Antigen Synonyms:
- Epithelial splicing regulatory protein 2|ESRP 2|ESRP2_HUMAN|FLJ21918|FLJ22248|RBM35B|RNA binding motif protein 35A|RNA binding motif protein 35B|RNA binding protein 35B|RNA-binding motif protein 35B|RNA-binding protein 35B
- Epithelial splicing regulatory protein 2|ESRP 2|ESRP2_HUMAN|FLJ21918|FLJ22248|RBM35B|RNA binding motif protein 35A|RNA binding motif protein 35B|RNA binding protein 35B|RNA-binding motif protein 35B|RNA-binding protein 35B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H6T0
- Q9H6T0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Epithelial splicing regulatory protein 2 protein (615-720AA)
- Recombinant Human Epithelial splicing regulatory protein 2 protein (615-720AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together