Anti-KRT13 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-851
- 77858-722
- Antigen Symbol:
- KRT13
- KRT13
- Antigen Synonyms:
- 47 kDa cytokeratin|CK-13|CK13|Cytokeratin 13|Cytokeratin-13|K13|K1C13_HUMAN|Ka13|Keratin 13|Keratin|keratin type I cytoskeletal 13|Keratin-13|Krt-1.13|Krt1-13|MGC161462|MGC3781|type I cytoskeletal 13|Type I keratin Ka13|WSN2
- 47 kDa cytokeratin|CK-13|CK13|Cytokeratin 13|Cytokeratin-13|K13|K1C13_HUMAN|Ka13|Keratin 13|Keratin|keratin type I cytoskeletal 13|Keratin-13|Krt-1.13|Krt1-13|MGC161462|MGC3781|type I cytoskeletal 13|Type I keratin Ka13|WSN2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13646
- P13646
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Keratin, type I cytoskeletal 13 protein (420-458AA)
- Recombinant Human Keratin, type I cytoskeletal 13 protein (420-458AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together