Anti-NMT1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NMT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-574
- 77744-573
- Antigen Symbol:
- NMT1
- NMT1
- Antigen Synonyms:
- Alternative, short form NMT S|EC 2.3.1.97|Glycylpeptide N tetradecanoyltransferase 1|Glycylpeptide N-tetradecanoyltransferase 1|Long form, NMT L|Myristoyl CoA:protein N myristoyltransferase 1|Myristoyl-CoA:protein N-myristoyltransferase 1|N myristoyltransferase 1|NMT 1|NMT|nmt1|NMT1_HUMAN|Peptide N myristoyltransferase 1|Peptide N-myristoyltransferase 1|Type I N myristoyltransferase|Type I N-myristoyltransferase
- Alternative, short form NMT S|EC 2.3.1.97|Glycylpeptide N tetradecanoyltransferase 1|Glycylpeptide N-tetradecanoyltransferase 1|Long form, NMT L|Myristoyl CoA:protein N myristoyltransferase 1|Myristoyl-CoA:protein N-myristoyltransferase 1|N myristoyltransferase 1|NMT 1|NMT|nmt1|NMT1_HUMAN|Peptide N myristoyltransferase 1|Peptide N-myristoyltransferase 1|Type I N myristoyltransferase|Type I N-myristoyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30419
- P30419
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Glycylpeptide N-tetradecanoyltransferase 1 protein (1-158AA)
- Glycylpeptide N-tetradecanoyltransferase 1 protein (1-158AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together