Anti-HMMR Rabbit Polyclonal Antibody
Catalog # 77737-943
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HMMR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O75330
- Antigen Synonyms:CD168|CD168 antigen|HMMR|HMMR_HUMAN|Hyaluronan mediated motility receptor|Hyaluronan-mediated motility receptor (RHAMM)|IHABP|Intracellular hyaluronic acid-binding protein|MGC119494|MGC119495|OTTHUMP00000196920|Receptor for hyaluronan-mediated motility|RHAMM
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:CD168
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, IHC:1:100 to 1:300, ELISA:1:40000
Type:
Antigen: HMMR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human