Anti-HCN1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HCN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-784
- 77739-544
- Antigen Symbol:
- HCN1
- HCN1
- Antigen Synonyms:
- BCNG-1|BCNG1|Brain cyclic nucleotide gated channel 1|Brain cyclic nucleotide-gated channel 1|HAC 2|HAC2|HCN1_HUMAN|Hyperpolarization activated cyclic nucleotide gated potassium channel 1|Potassium channel|voltage-gated|brain|1|Potassium/sodium hyperpolarization activated cyclic nucleotide gated channel 1|Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
- BCNG-1|BCNG1|Brain cyclic nucleotide gated channel 1|Brain cyclic nucleotide-gated channel 1|HAC 2|HAC2|HCN1_HUMAN|Hyperpolarization activated cyclic nucleotide gated potassium channel 1|Potassium channel|voltage-gated|brain|1|Potassium/sodium hyperpolarization activated cyclic nucleotide gated channel 1|Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60741
- O60741
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1 protein (784-887AA)
- Recombinant Human Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1 protein (784-887AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together