Anti-Gamma-glutamyl hydrolase Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: Gamma-glutamyl hydrolase
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Glycine max
- Catalog No:
- 77855-955
- 77738-203
- Antigen Symbol:
- Gamma-glutamyl hydrolase
- Gamma-glutamyl hydrolase
- Antigen Synonyms:
- Gamma-glutamyl hydrolase| EC 3.4.19.9| Conjugase| GH| Gamma-Glu-X carboxypeptidase
- Gamma-glutamyl hydrolase|EC 3.4.19.9|Conjugase|GH|Gamma-Glu-X carboxypeptidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Glycine max
- Glycine max
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P93164
- P93164
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Glycine max Gamma-glutamyl hydrolase protein (22-342AA)
- Recombinant Glycine max Gamma-glutamyl hydrolase protein (22-342AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together