Anti-HARS Rabbit Polyclonal Antibody
Catalog # 77739-418
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HARS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P12081
- Antigen Synonyms:cytoplasmic| EC 6.1.1.21| FLJ20491| HisRS| Histidine tRNA ligase, cytoplasmic| histidine translase| Histidine tRNA ligase| Histidine--tRNA ligase| Histidyl tRNA synthetase| Histidyl-tRNA synthetase| HRS| Human histidyl tRNA synthetase homolog (HO3) mRNA complete cds| SYHC_HUMAN| USH3B
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:USH3B
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat