Anti-GP9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GP9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77738-699
- 77856-277
- Antigen Symbol:
- GP9
- GP9
- Antigen Synonyms:
- CD42a|Glycoprotein 9|Glycoprotein IX|Glycoprotein IX (platelet)|Glycoprotein IX precursor|GP IX|GP-IX|GPIX|GPIX_HUMAN|Platelet glycoprotein IX
- CD42a|Glycoprotein 9|Glycoprotein IX|Glycoprotein IX (platelet)|Glycoprotein IX precursor|GP IX|GP-IX|GPIX|GPIX_HUMAN|Platelet glycoprotein IX
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14770
- P14770
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Platelet glycoprotein IX protein (17-88AA)
- Recombinant Human Platelet glycoprotein IX protein (17-88AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together