Anti-HNRNPC Rabbit Polyclonal Antibody
Catalog # 77740-723
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HNRNPC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P07910
- Antigen Synonyms:C1|C2|Heterogeneous nuclear ribonucleoprotein C (C1/C2)|Heterogeneous nuclear ribonucleoprotein C|Heterogeneous nuclear ribonucleoproteins C1/C2|HNRNP|hnRNP C1 / hnRNP C2|hnRNP C1/C2|Hnrnpc|HNRPC|HNRPC_HUMAN|MGC104306|MGC105117|MGC117353|MGC131677|Nuclear ribonucleoprotein particle C1 protein|Nuclear ribonucleoprotein particle C2 protein|SNRPC
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:C1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: HNRNPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat