Anti-NRG2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NRG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-033
- 77745-034
- Antigen Symbol:
- NRG2
- NRG2
- Antigen Synonyms:
- Divergent of neuregulin 1|Divergent of neuregulin-1|Don 1|DON-1|Neural- and thymus-derived activator for ERBB kinases|Neuregulin 2|Neuregulin-2|NRG-2|Nrg2|NRG2_HUMAN|NTAK|Pro NRG2|Pro-NRG2
- Divergent of neuregulin 1|Divergent of neuregulin-1|Don 1|DON-1|Neural- and thymus-derived activator for ERBB kinases|Neuregulin 2|Neuregulin-2|NRG-2|Nrg2|NRG2_HUMAN|NTAK|Pro NRG2|Pro-NRG2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14511
- O14511
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Pro-neuregulin-2, membrane-bound isoform protein (112-405AA)
- Pro-neuregulin-2, membrane-bound isoform protein (112-405AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together