Anti-IARS2 Rabbit Polyclonal Antibody
Catalog # 77740-552
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NSE4
- Antigen Synonyms:2010002H18Rik|C79125|EC 6.1.1.5|FLJ10326|Iars2|IleRS|isoleucine tRNA ligase 2, mitochondrial|isoleucine tRNA ligase, mitochondrial|isoleucine tRNA synthetase 2, mitochondrial|Isoleucine--tRNA ligase|isoleucine-tRNA synthetase 2|isoleucyl tRNA synthetase 2, mitochondrial|Isoleucyl-tRNA synthetase|MGC63429|mitochondrial|mitochondrial isoleucine tRNA synthetase|rCG_20084|RGD1311857|SYIM_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:SYIM_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat