Anti-MIP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MIP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-149
- 77859-768
- Antigen Symbol:
- MIP
- MIP
- Antigen Synonyms:
- AQP 0|AQP0|Aquaporin|Aquaporin 0|Aquaporin-0|Aquaporin0|Lens fiber major intrinsic protein|LIM 1|LIM1|Major intrinsic protein 26kD|Major intrinsic protein of lens fiber|MIP 26|MIP_HUMAN|MIP26|MP 26|MP26|PALM
- AQP 0|AQP0|Aquaporin|Aquaporin 0|Aquaporin-0|Aquaporin0|Lens fiber major intrinsic protein|LIM 1|LIM1|Major intrinsic protein 26kD|Major intrinsic protein of lens fiber|MIP 26|MIP_HUMAN|MIP26|MP 26|MP26|PALM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30301
- P30301
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lens fiber major intrinsic protein (220-263AA)
- Recombinant Human Lens fiber major intrinsic protein (220-263AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together