Anti-MYNN Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: MYNN
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-767
- 77860-217
- Antigen Symbol:
- MYNN
- MYNN
- Antigen Synonyms:
- MYNN_HUMAN| Myoneurin| OSZF| SBBIZ1| ZBTB31| Zinc finger and BTB domain containing protein 31| Zinc finger and BTB domain-containing protein 31| Zinc finger protein with BTB/POZ domain| ZNF902
- MYNN_HUMAN|Myoneurin|OSZF|SBBIZ1|ZBTB31|Zinc finger and BTB domain containing protein 31|Zinc finger and BTB domain-containing protein 31|Zinc finger protein with BTB/POZ domain|ZNF902
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPC7
- Q9NPC7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Myoneurin protein (51-330AA)
- Recombinant Human Myoneurin protein (51-330AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together