Anti-MAP4K4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP4K4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-431
- 77742-713
- Antigen Symbol:
- MAP4K4
- MAP4K4
- Antigen Synonyms:
- FLH21957|FLJ10410|FLJ20373|FLJ90111|Hepatocyte progenitor kinase like/germinal center kinase like kinase|HGK|HPK/GCK like kinase|HPK/GCK like kinase HGK|HPK/GCK-like kinase HGK|KIAA0687|M4K4_HUMAN|MAPK/ERK kinase kinase kinase 4|MEK kinase kinase 4|MEKKK 4|MEKKK4|Mitogen activated protein kinase kinase kinase kinase 4|Mitogen-activated protein kinase kinase kinase kinase 4|Nck interacting kinase|Nck-interacting kinase|NIK
- FLH21957|FLJ10410|FLJ20373|FLJ90111|Hepatocyte progenitor kinase like/germinal center kinase like kinase|HGK|HPK/GCK like kinase|HPK/GCK like kinase HGK|HPK/GCK-like kinase HGK|KIAA0687|M4K4_HUMAN|MAPK/ERK kinase kinase kinase 4|MEK kinase kinase 4|MEKKK 4|MEKKK4|Mitogen activated protein kinase kinase kinase kinase 4|Mitogen-activated protein kinase kinase kinase kinase 4|Nck interacting kinase|Nck-interacting kinase|NIK
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95819
- O95819
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Mitogen-activated protein kinase kinase kinase kinase 4 protein (866-1104AA)
- Recombinant human Mitogen-activated protein kinase kinase kinase kinase 4 protein (866-1104AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together