Anti-HEXB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HEXB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-655
- 77856-832
- Antigen Symbol:
- HEXB
- HEXB
- Antigen Synonyms:
- Beta hexosaminidase beta chain|Beta hexosaminidase subunit beta|Beta N acetylhexosaminidase|Beta-hexosaminidase subunit beta chain A|Beta-N-acetylhexosaminidase subunit beta|Cervical cancer proto oncogene 7 protein|Cervical cancer proto-oncogene 7 protein|ENC 1AS|Epididymis luminal protein 248|HCC 7|HCC-7|HCC7|HEL 248|HEX B|HEXB_HUMAN|Hexosaminidase B (beta polypeptide)|Hexosaminidase B|Hexosaminidase subunit B|HexosaminidaseB|N acetyl beta glucosaminidase|N-acetyl-beta-glucosaminidase subunit beta
- Beta hexosaminidase beta chain|Beta hexosaminidase subunit beta|Beta N acetylhexosaminidase|Beta-hexosaminidase subunit beta chain A|Beta-N-acetylhexosaminidase subunit beta|Cervical cancer proto oncogene 7 protein|Cervical cancer proto-oncogene 7 protein|ENC 1AS|Epididymis luminal protein 248|HCC 7|HCC-7|HCC7|HEL 248|HEX B|HEXB_HUMAN|Hexosaminidase B (beta polypeptide)|Hexosaminidase B|Hexosaminidase subunit B|HexosaminidaseB|N acetyl beta glucosaminidase|N-acetyl-beta-glucosaminidase subunit beta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07686
- P07686
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Beta-hexosaminidase subunit beta protein (408-528AA)
- Recombinant Human Beta-hexosaminidase subunit beta protein (408-528AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together