Anti-MCEE Rabbit Polyclonal Antibody
Catalog # 77742-880
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MCEE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96PE7
- Antigen Synonyms:DL methylmalonyl CoA racemase| DL-methylmalonyl-CoA racemase| EC 5.1.99.1| GLOD2 | Glyoxalase domain containing 2 | MCEE_HUMAN| Methylmalonyl CoA epimerase| Methylmalonyl-CoA epimerase| methylmalonyl-CoA epimerase| mitochondrial| OTTHUMP00000160122
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:OTTHUMP00000160122
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MCEE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat