Anti-FSD1L Rabbit Polyclonal Antibody
Catalog # 77737-662
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FSD1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BXM9
- Antigen Synonyms:CCDC10|CSDUFD1|FSD1CL|FSD1NLFSD1-like protein|Coiled-coil domain-containing protein 10|FSD1 N-terminal-like protein
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:FSD1 N-terminal-like protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FSD1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat