Anti-FUT3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: FUT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77737-728
- 77855-654
- Antigen Symbol:
- FUT3
- FUT3
- Antigen Synonyms:
- Alpha-(1|3/1|4)-fucosyltransferase|Blood group Lewis alpha-4-fucosyltransferase|CD174|FT3B|Fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase|Lewis blood group)|Fucosyltransferase 3|Fucosyltransferase III|FucT III|FucT-III|FUT3_HUMAN|Galactoside 3(4)-L-fucosyltransferase|LE|Les|Lewis FT
- Alpha-(1|3/1|4)-fucosyltransferase|Blood group Lewis alpha-4-fucosyltransferase|CD174|FT3B|Fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase|Lewis blood group)|Fucosyltransferase 3|Fucosyltransferase III|FucT III|FucT-III|FUT3_HUMAN|Galactoside 3(4)-L-fucosyltransferase|LE|Les|Lewis FT
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21217
- P21217
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Galactoside 3(4)-L-fucosyltransferase protein (83-187AA)
- Recombinant Human Galactoside 3(4)-L-fucosyltransferase protein (83-187AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together