Anti-MKNK1 Rabbit Polyclonal Antibody
Catalog # 77740-188
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MKNK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Form:Liquid
- Gene ID:Q9BUB5
- Antigen Synonyms:MAP kinase interacting kinase 1|MAP kinase interacting serine/threonine kinase 1|MAP kinase signal integrating kinase 1|MAP kinase signal-integrating kinase 1|MAP kinase-interacting serine/threonine-protein kinase 1|MAPK signal integrating kinase 1|MITOGEN-ACTIVATED PROTEIN KINASE-INTERACTING SERINE/THREONINE KINASE 1|mknk1|MKNK1_HUMAN|MNK 1|Mnk1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MAP kinase interacting kinase 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:100 to 1:300, ELISA:1:20000
Type:
Antigen: MKNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat