Anti-NR3C1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NR3C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-973
- 77744-974
- Antigen Symbol:
- NR3C1
- NR3C1
- Antigen Synonyms:
- GCCR|GCR|GCR_HUMAN|GCRST|glucocorticoid nuclear receptor variant 1|Glucocorticoid receptor|GR|GRL|Grl1|Nuclear receptor subfamily 3 group C member 1|nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
- GCCR|GCR|GCR_HUMAN|GCRST|glucocorticoid nuclear receptor variant 1|Glucocorticoid receptor|GR|GRL|Grl1|Nuclear receptor subfamily 3 group C member 1|nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04150
- P04150
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Glucocorticoid receptor protein (1-190AA)
- Glucocorticoid receptor protein (1-190AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together