Anti-ERLIN1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ERLIN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-129
- 77854-898
- Antigen Symbol:
- ERLIN1
- ERLIN1
- Antigen Synonyms:
- Band 7 23-211 Keo4 (Interim) similar to C.elegans protein|C10orf69|chromosome 10 open reading frame 69|Endoplasmic reticulum lipid raft-associated protein 1|ER lipid raft associated 1|Erlin 1|Erlin-1|ERLN1_HUMAN|KE04|KEO4|OTTHUMP00000059425|Protein KE04|SPFH domain containing protein 1|SPFH domain family|member 1|SPFH domain-containing protein 1|SPFH1|SPG62|Stomatin-prohibitin-flotillin-HflC/K domain containing protein 1|Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
- Band 7 23-211 Keo4 (Interim) similar to C.elegans protein|C10orf69|chromosome 10 open reading frame 69|Endoplasmic reticulum lipid raft-associated protein 1|ER lipid raft associated 1|Erlin 1|Erlin-1|ERLN1_HUMAN|KE04|KEO4|OTTHUMP00000059425|Protein KE04|SPFH domain containing protein 1|SPFH domain family|member 1|SPFH domain-containing protein 1|SPFH1|SPG62|Stomatin-prohibitin-flotillin-HflC/K domain containing protein 1|Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75477
- O75477
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Erlin-1 protein (27-346AA)
- Recombinant Human Erlin-1 protein (27-346AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together