Anti-EPRS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: EPRS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77736-053
- 77854-855
- Antigen Symbol:
- EPRS
- Antigen Synonyms:
- Bifunctional aminoacyl tRNA synthetase|Bifunctional aminoacyl-tRNA synthetase|Bifunctional glutamate/proline tRNA ligase|Cell proliferation-inducing gene 32 protein|DKFZp313B047|EARS|Eprs|GLNS|Glu pro tRNA synthetase|GLUPRORS|GluRS|Glutamate tRNA ligase|Glutamatyl prolyl tRNA synthetase|Glutaminyl tRNA synthetase|Glutamyl prolyl tRNA synthetase|Glutamyl tRNA synthetase|Glutamyl-tRNA synthetase|PARS|PIG 32|PIG32|Proliferation inducing gene 32 protein|Proliferation inducing protein 32|Proline tRNA ligase|Proline--tRNA ligase|Prolyl tRNA synthetase|Prolyl-tRNA synthetase|QARS|QPRS|SYEP_HUMAN
- Bifunctional aminoacyl tRNA synthetase|Bifunctional aminoacyl-tRNA synthetase|Bifunctional glutamate/proline tRNA ligase|Cell proliferation-inducing gene 32 protein|DKFZp313B047|EARS|Eprs|GLNS|Glu pro tRNA synthetase|GLUPRORS|GluRS|Glutamate tRNA ligase|Glutamatyl prolyl tRNA synthetase|Glutaminyl tRNA synthetase|Glutamyl prolyl tRNA synthetase|Glutamyl tRNA synthetase|Glutamyl-tRNA synthetase|PARS|PIG 32|PIG32|Proliferation inducing gene 32 protein|Proliferation inducing protein 32|Proline tRNA ligase|Proline--tRNA ligase|Prolyl tRNA synthetase|Prolyl-tRNA synthetase|QARS|QPRS|SYEP_HUMAN
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P07814
- P07814
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Bifunctional glutamate/proline--tRNA ligase protein (1207-1399AA)
- Recombinant Human Bifunctional glutamate/proline--tRNA ligase protein (1207-1399AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together