Anti-GPR183 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPR183
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-839
- 77856-345
- Antigen Symbol:
- GPR183
- GPR183
- Antigen Synonyms:
- ebi 2|EBI2|EBV induced G protein coupled receptor 2|EBV-induced G-protein coupled receptor 2|ebv-induced gene 2|Epstein Barr virus induced gene 2 (lymphocyte specific G protein coupled receptor)|Epstein Barr virus induced gene 2|epstein-barr virus induced gene 2|Epstein-Barr virus-induced G-protein coupled receptor 2|G protein-coupled receptor 183|G-protein coupled receptor 183|GP183_HUMAN|gpr183|hEBI2|Lymphocyte specific G protein coupled receptor
- ebi 2| EBI2| EBV induced G protein coupled receptor 2| EBV-induced G-protein coupled receptor 2| ebv-induced gene 2| Epstein Barr virus induced gene 2 (lymphocyte specific G protein coupled receptor)| Epstein Barr virus induced gene 2| epstein-barr virus induced gene 2| Epstein-Barr virus-induced G-protein coupled receptor 2| G protein-coupled receptor 183| G-protein coupled receptor 183| GP183_HUMAN| gpr183| hEBI2| Lymphocyte specific G protein coupled receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32249
- P32249
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human G-protein coupled receptor 183 protein (1-31AA)
- Recombinant Human G-protein coupled receptor 183 protein (1-31AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together