Anti-MGAT3 Rabbit Polyclonal Antibody
Catalog # 77743-087
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MGAT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q09327
- Antigen Synonyms:4-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase|Beta 1 4 mannosyl glycoprotein 4 beta N acetylglucosaminyltransferase|Beta-1|FLJ43371|GlcNAc T III|GlcNAc-T III|GNT 3|GNT III|GNT-III|GNT3|Mannosyl (beta 1 4 ) glycoprotein beta 1 4 N acetylglucosaminyltransferase|MGAT 3|MGAT3_HUMAN|MGC141943|MGC142278|N acetylglucosaminyltransferase III|N glycosyl oligosaccharide glycoprotein N acetylglucosaminyltransferase III|N-acetylglucosaminyltransferase III|N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase III|OTTHUMP00000042207
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:OTTHUMP00000042207
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MGAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat