Anti-LAT2 Rabbit Polyclonal Antibody
Catalog # 77741-997
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9GZY6
- Antigen Synonyms:HSPC046|LAB|Linker for activation of B cells|Linker for activation of B-cells|Linker for activation of T cells family member 2|Linker for activation of T cells transmembrane adaptor 2|Linker for activation of T-cells family member 2|Membrane associated adapter molecule|Membrane-associated adapter molecule|Non T cell activation linker|Non-T-cell activation linker|NTAL_HUMAN|WBSCR 5|Wbscr15|Williams Beuren syndrome chromosome region 15|Williams Beuren syndrome chromosome region 5|Williams-Beuren syndrome chromosomal region 15 protein|Williams-Beuren syndrome chromosomal region 5 protein|WSCR5
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:WSCR5
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: LAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human