Anti-LANCL1 Rabbit Polyclonal Antibody
Catalog # 77741-978
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LANCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O43813
- Antigen Synonyms:40 kDa erythrocyte membrane protein|G protein-coupled receptor 69A|GPR69A|LanC (bacterial lantibiotic synthetase component C) like 1|LanC lantibiotic synthetase component C like 1 (bacterial)|LanC-like protein 1|LANC1_HUMAN|p40
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:p40
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: LANCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat