Anti-MSC Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: MSC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-020
- 77743-506
- Antigen Symbol:
- MSC
- MSC
- Antigen Synonyms:
- ABF-1| ABF1| Activated B cell factor 1| Activated B-cell factor 1| bHLHa22| Class A basic helix-loop-helix protein 22| Msc| MUSC_HUMAN| Musculin| MYOR
- ABF-1|ABF1|Activated B cell factor 1|Activated B-cell factor 1|bHLHa22|Class A basic helix-loop-helix protein 22|Msc|MUSC_HUMAN|Musculin|MYOR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60682
- O60682
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Musculin protein (3-143AA)
- Recombinant Human Musculin protein (3-143AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together