Anti-NF1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: NF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-340
- 77744-339
- Antigen Symbol:
- NF1
- NF1
- Antigen Synonyms:
- DKFZp686J1293|FLJ21220|Neurofibromatosis Noonan syndrome|Neurofibromatosis related protein NF 1|Neurofibromatosis related protein NF1|neurofibromatosis type I|Neurofibromatosis-related protein NF-1|Neurofibromin 1|Neurofibromin truncated|Neurofibromin1|NF 1|NF|NF1_HUMAN|NFNS|Type 1 Neurofibromatosis|von Recklinghausen disease neurofibromin|von Recklinghausen disease related protein VRNF|VRNF|WATS|Watson disease related protein WSS|Watson syndrome|WSS
- DKFZp686J1293|FLJ21220|Neurofibromatosis Noonan syndrome|Neurofibromatosis related protein NF 1|Neurofibromatosis related protein NF1|neurofibromatosis type I|Neurofibromatosis-related protein NF-1|Neurofibromin 1|Neurofibromin truncated|Neurofibromin1|NF 1|NF|NF1_HUMAN|NFNS|Type 1 Neurofibromatosis|von Recklinghausen disease neurofibromin|von Recklinghausen disease related protein VRNF|VRNF|WATS|Watson disease related protein WSS|Watson syndrome|WSS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21359
- P21359
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Neurofibromin protein (655-892AA)
- Neurofibromin protein (655-892AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together