Anti-IL10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Macaca mulatta
Specifications
- Catalog No:
- 77740-924
- 77857-995
- Antigen Symbol:
- IL10
- IL10
- Antigen Synonyms:
- IL10Interleukin-10|IL-10|Cytokine synthesis inhibitory factor|CSIF
- IL10Interleukin-10| IL-10| Cytokine synthesis inhibitory factor| CSIF
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Macaca mulatta
- Macaca mulatta
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51496
- P51496
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Macaca mulatta Interleukin-10 protein (19-178AA)
- Recombinant Macaca mulatta Interleukin-10 protein (19-178AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together