Anti-NCAM2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NCAM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-976
- 77743-977
- Antigen Symbol:
- NCAM2
- NCAM2
- Antigen Synonyms:
- Cell adhesion molecule|neural|2|MGC51008|N CAM 2|N CAM 21|N CAM2|N CAM21|N-CAM-2|NCAM 2|NCAM 21|NCAM-2|Ncam2|NCAM2_HUMAN|NCAM21|Neural cell adhesion molecule 2|OCAM GPI|R4B12|RB 8 neural cell adhesion molecule|RB8 neural cell adhesion molecule|RNCAM
- Cell adhesion molecule|neural|2|MGC51008|N CAM 2|N CAM 21|N CAM2|N CAM21|N-CAM-2|NCAM 2|NCAM 21|NCAM-2|Ncam2|NCAM2_HUMAN|NCAM21|Neural cell adhesion molecule 2|OCAM GPI|R4B12|RB 8 neural cell adhesion molecule|RB8 neural cell adhesion molecule|RNCAM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15394
- O15394
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Neural cell adhesion molecule 2 protein (209-333AA)
- Neural cell adhesion molecule 2 protein (209-333AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together