Anti-KLRC2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KLRC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-676
- 77741-795
- Antigen Symbol:
- KLRC2
- KLRC2
- Antigen Synonyms:
- CD antigen CD159c|CD159 antigen like family member C|CD159 antigen-like family member C|CD159c|Killer cell lectin like receptor subfamily C, member 2|NK cell receptor C|NKG2-C type II integral membrane protein|NKG2-C-activating NK receptor|NKG2C activating NK receptor|NKG2C|NKG2C_HUMAN
- CD antigen CD159c|CD159 antigen like family member C|CD159 antigen-like family member C|CD159c|Killer cell lectin like receptor subfamily C, member 2|NK cell receptor C|NKG2-C type II integral membrane protein|NKG2-C-activating NK receptor|NKG2C activating NK receptor|NKG2C|NKG2C_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P26717
- P26717
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human NKG2-C type II integral membrane protein (94-231AA)
- Recombinant human NKG2-C type II integral membrane protein (94-231AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together