Anti-GPR17 Rabbit Polyclonal Antibody
Catalog # 77738-827
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GPR17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13304
- Antigen Synonyms:DKFZp686M18273|G protein coupled receptor 17|G-protein coupled receptor 17|GPR 17|GPR17_HUMAN|P2Y like receptor|P2Y-like receptor|Probable P2Y purinoceptor GPR 17|Probable P2Y purinoceptor R12|UDP/CysLT receptor|Uracil nucleotide/cysteinyl leukotriene receptor
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Uracil nucleotide/cysteinyl leukotriene receptor
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GPR17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat