Order Entry
United States
ContactUsLinkComponent
Anti-HSPA12A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-HSPA12A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-HSPA12A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77740-457
  • 77857-704
  • Antigen Symbol:
  • HSPA12A
  • HSPA12A
  • Antigen Synonyms:
  • FLJ13874|Heat shock 70 kDa protein 12A|heat shock 70kD protein 12A|heat shock 70kDa protein 12A|HS12A_HUMAN|Hspa12a|KIAA0417
  • FLJ13874|Heat shock 70 kDa protein 12A|heat shock 70kD protein 12A|heat shock 70kDa protein 12A|HS12A_HUMAN|Hspa12a|KIAA0417
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O43301
  • O43301
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Heat shock 70 kDa protein 12A protein (2-231AA)
  • Recombinant Human Heat shock 70 kDa protein 12A protein (2-231AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together