Anti-GPLD1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: GPLD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77856-318
- 77738-758
- Antigen Symbol:
- GPLD1
- GPLD1
- Antigen Synonyms:
- Glycoprotein phospholipase D|Glycosyl-phosphatidylinositol-specific phospholipase D|GPI-PLD|GPI-specific phospholipase D|GPIPLD|GPIPLDM|MGC22590|PHLD_HUMAN|Phosphatidylinositol glycan specific phospholipase D|Phosphatidylinositol-glycan-specific phospholipase D|PI-G PLD|PIGPLD|PIGPLD1
- Glycoprotein phospholipase D|Glycosyl-phosphatidylinositol-specific phospholipase D|GPI-PLD|GPI-specific phospholipase D|GPIPLD|GPIPLDM|MGC22590|PHLD_HUMAN|Phosphatidylinositol glycan specific phospholipase D|Phosphatidylinositol-glycan-specific phospholipase D|PI-G PLD|PIGPLD|PIGPLD1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P80108
- P80108
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Phosphatidylinositol-glycan-specific phospholipase D protein (310-565AA)
- Recombinant Human Phosphatidylinositol-glycan-specific phospholipase D protein (310-565AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together