Anti-CERS1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CERS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-992
- 77858-839
- Antigen Symbol:
- CERS1
- CERS1
- Antigen Synonyms:
- Ceramide synthase 1|CerS1|CERS1_HUMAN|LAG1|LAG1 homolog ceramide synthase 1|LAG1 longevity assurance homolog 1|LASS 1|LASS1|Longevity assurance (LAG1 S. cerevisiae) homolog 1|Longevity assurance gene 1|Longevity assurance gene 1 protein homolog 1|MGC90349|Protein LAG1|Protein UOG 1|Protein UOG-1|UOG 1 protein|UOG1|Upstream of GDF1
- Ceramide synthase 1|CerS1|CERS1_HUMAN|LAG1|LAG1 homolog ceramide synthase 1|LAG1 longevity assurance homolog 1|LASS 1|LASS1|Longevity assurance (LAG1 S. cerevisiae) homolog 1|Longevity assurance gene 1|Longevity assurance gene 1 protein homolog 1|MGC90349|Protein LAG1|Protein UOG 1|Protein UOG-1|UOG 1 protein|UOG1|Upstream of GDF1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27544
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ceramide synthase 1 protein (1-52AA)
- Recombinant Human Ceramide synthase 1 protein (1-52AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together