Anti-CNMD Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CNMD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-060
- 77858-890
- Antigen Symbol:
- CNMD
- Antigen Synonyms:
- BRICD3|BRICHOS domain containing 3|CH-SP|CHM I|ChM-I|CHM1|chondromodulin|Chondromodulin I|Chondromodulin-1|Chondromodulin-I|Chondrosurfactant protein|LECT 1|Lect1|LECT1_HUMAN|Leukocyte cell derived chemotaxin 1|Multiple myeloma tumor suppressor 1|MYETS1
- BRICD3|BRICHOS domain containing 3|CH-SP|CHM I|ChM-I|CHM1|chondromodulin|Chondromodulin I|Chondromodulin-1|Chondromodulin-I|Chondrosurfactant protein|LECT 1|Lect1|LECT1_HUMAN|Leukocyte cell derived chemotaxin 1|Multiple myeloma tumor suppressor 1|MYETS1
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O75829
- O75829
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Leukocyte cell-derived chemotaxin 1 protein (215-334AA)
- Recombinant Human Leukocyte cell-derived chemotaxin 1 protein (215-334AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together