Anti-FOSB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: FOSB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-381
- 77855-519
- Antigen Symbol:
- FOSB
- FOSB
- Antigen Synonyms:
- Activator protein 1|AP 1|DKFZp686C0818|FBJ murine osteosarcoma viral oncogene homolog B|FOSB_HUMAN|G0/G1 switch regulatory protein 3|G0S3|GOS 3|GOS B|GOS3|GOSB|MGC42291|Oncogene FOS B|Oncogene Protein fos B|Protein fosB
- Activator protein 1|AP 1|DKFZp686C0818|FBJ murine osteosarcoma viral oncogene homolog B|FOSB_HUMAN|G0/G1 switch regulatory protein 3|G0S3|GOS 3|GOS B|GOS3|GOSB|MGC42291|Oncogene FOS B|Oncogene Protein fos B|Protein fosB
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53539
- P53539
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein fosB protein (1-41AA)
- Recombinant Human Protein fosB protein (1-41AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together